1,2-Dichlorobenzene-[D₄] ≥99% (Isotopic)
Supplier: BeanTown Chemical
Synonyms:
1,2-Dichlorobenzene-[²H₄]
220325-1G
220325-5G
BT220325-1GEA
90
USD
BT220325-1G
BT220325-5G
1,2-Dichlorobenzene-[D₄] ≥99% (Isotopic)
1,2-Dichlorobenzene-[D₄]
CAS: 2199-69-1; EC No: 218-606-0; MDL No: MFCD00037106
UN No: UN1591; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6D4Cl2; MW: 151.03
Boiling Point: 178-180°; Flash point: 66°C (151°F)
Density (g/mL): 1.341; Refractive Index: 1.551
- Refractive index: 1.551
- Liquid
Formula:
C₆D₄Cl₂ MW: 150.97 g/mol Boiling Pt: 178…180 °C Density: 1.341 g/mL Flash Pt: 66 °C (151 °F) |
MDL Number:
MFCD00037106 CAS Number: 2199-69-1 EINECS: 218-606-0 UN: 1591 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...