Calcium oxide

Supplier: BeanTown Chemical

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

220310-2KG 220310-500G
BT220310-2KGEA 272.09 USD
BT220310-2KG BT220310-500G
Calcium oxide
Calcium oxide

CAS: 1305-78-8; EC No: 215-138-9; MDL No: MFCD00010911 UN No: UN1910; Haz Class: 8; Packing Group: III Powder; Molecular Formula: CaO; MW: 56.08 Boiling Point: 2850° Density (g/mL): 3.3 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: CaO
MW: 56.08 g/mol
Boiling Pt: 2850 °C
Density: 3.3 g/mL
MDL Number: MFCD00010911
CAS Number: 1305-78-8
EINECS: 215-138-9
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR