Potassium-tert-pentyloxide 14 - 18% (w/v) in cyclohexane

Supplier: BeanTown Chemical

Synonyms: Potassium tert-amylate, Potassium 2-methyl-2-butoxide, Potassium tert-pentoxide

220270-100G 220270-25G 220270-5G
BT220270-100GEA 307 USD
BT220270-100G BT220270-25G BT220270-5G
Potassium-tert-pentyloxide 14 - 18% (w/v) in cyclohexane
Potassium-tert-pentyloxide

CAS: 41233-93-6; EC No: 255-272-5; MDL No: MFCD00064808 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Molecular Formula: C5H11KO; MW: 126.24 Flash point: -18°C (0°F) Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: C₅H₁₁KO
MW: 126.24 g/mol
Flash Pt: –18 °C (0 °F)
MDL Number: MFCD00064808
CAS Number: 41233-93-6
EINECS: 255-272-5
ADR: 3(8),II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR