2,4-Difluoroaniline ≥98%
Supplier: BeanTown Chemical
220225-100G
220225-25G
220225-500G
BT220225-100GEA
173
USD
BT220225-100G
BT220225-25G
BT220225-500G
2,4-Difluoroaniline ≥98%
2,4-Difluoroaniline
CAS: 367-25-9; EC No: 206-687-5; MDL No: MFCD00007648; RTECS: BX3680000
UN No: UN2941 ; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C6H5F2N ; MW: 129.11
Melting Point: -7.5°; Boiling Point: 170°/753 mmHg; Flash point: 66°C (144°F)
Density (g/mL): 1.268; Refractive Index: 1.506
- RTECS: BX3680000
- Refractive index: 1.506
- Liquid
Formula:
C₆H₅F₂N MW: 129.11 g/mol Boiling Pt: 170 °C/753 mmHg Melting Pt: –7.5 °C Density: 1.268 g/mL Flash Pt: 66 °C (144 °F) |
MDL Number:
MFCD00007648 CAS Number: 367-25-9 EINECS: 206-687-5 UN: 2941 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...