Lutetium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Supplier: BeanTown Chemical
220215-25X25MM
220215-50X50MM
BT220215-25X25EA
454
USD
BT220215-25X25
BT220215-50X50
Lutetium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.1 mm
Lutetium
CAS: 7439-94-3; EC No: 231-103-0; MDL No: MFCD00011098
UN No: UN3178; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: Lu; MW: 174.97
Melting Point: 1663°; Boiling Point: 3402°
Density (g/mL): 9.84
- Solid
Formula:
Lu MW: 174.97 g/mol Boiling Pt: 3402 °C Melting Pt: 1663 °C Density: 9.84 g/mL |
MDL Number:
MFCD00011098 CAS Number: 7439-94-3 EINECS: 231-103-0 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...