Adipoyl dichloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Adipoyl chloride

220070-100G 220070-25G 220070-500G
BT220070-100GEA 145 USD
BT220070-100G BT220070-25G BT220070-500G
Adipoyl dichloride ≥98%
Adipoyl dichloride

CAS: 111-50-2; EC No: 203-876-4; MDL No: MFCD00000759 UN No: UN3265; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C6H8Cl2O2; MW: 183.03 Boiling Point: 105-107°/2 mmHg; Flash point: 112°C (234°F) Density (g/mL): 1.259; Refractive Index: 1.471 Moisture Sensitive


  • Refractive index: 1.471
  • Moisture Sensitive
  • Liquid

Formula: C₆H₈Cl₂O₂
Boiling Pt: 105…107 °C/2 mmHg
Density: 1.259 g/mL
Flash Pt: 112 °C (234 °F)
MDL Number: MFCD00000759
CAS Number: 111-50-2
EINECS: 203-876-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR