Methotrexate

Supplier: BeanTown Chemical

Synonyms: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid

220060-100MG 220060-1G
BT220060-100MGEA 124 USD
BT220060-100MG BT220060-1G
Methotrexate
Methotrexate

CAS: 59-05-2; EC No: 200-413-8; MDL No: MFCD00150847; RTECS: MA1225000 UN No: UN1544; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C20H22N8O5; MW: 454.44 Melting Point: 185-204°


  • RTECS: MA1225000
  • Powder

Formula: C₂₀H₂₂N₈O₅
MW: 454.44 g/mol
Melting Pt: 185…204 °C
MDL Number: MFCD00150847
CAS Number: 59-05-2
EINECS: 200-413-8
UN: 1544
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR