Ishikawa's reagent ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
1-(Diethylamino)-1,1,2,3,3,3-hexafluoropropane, Hexafluoropropene diethylamine, N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine
219935-100G
219935-25G
BT219935-100GEA
213
USD
BT219935-100G
BT219935-25G
Ishikawa's reagent ≥90%, Technical Grade
Ishikawa's reagent
CAS: 309-88-6; MDL No: MFCD00054683
UN No: UN2924; Haz Class: 3(8); Packing Group: II
Liquid; Molecular Formula: C7H11F6N; MW: 223.16
Boiling Point: 56-57°; Flash point: 14°C (57°F)
Density (g/mL): 1.230
- Liquid
Formula:
C₇H₁₁F₆N MW: 223.16 g/mol Boiling Pt: 56…57 °C Density: 1.230 g/mL Flash Pt: 14 °C (57 °F) |
MDL Number:
MFCD00054683 CAS Number: 309-88-6 ADR: 3(8),II |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...