Hydrogen peroxide 35% (w/w) in water stabilised
Supplier: BeanTown Chemical
219920-100ML
219920-500ML
BT219920-100MLEA
33.55
USD
BT219920-100ML
BT219920-500ML
Hydrogen peroxide 35% (w/w) in water stabilised
Hydrogen peroxide
CAS: 7722-84-1; EC No: 231-765-0; MDL No: MFCD00011333; RTECS: MX0899000
UN No: UN2014; Haz Class: 5.1 (8); Packing Group: II
Liquid; Molecular Formula: H2O2; MW: 34.02
- RTECS: MX0899000
- Liquid
Formula:
H₂O₂ MW: 34.01 Boiling Pt: 150.2 Melting Pt: -0.41 Density: 1.11 |
MDL Number:
MFCD00011333 CAS Number: 7722-84-1 EINECS: 231-765-0 UN: 2014 ADR: 5.1 II Merck Index: 14,04798 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...