Tin(II) iodide, anhydrous ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Tin(II) iodide, Tin diiodide

ampouled under argon

219915-25G 219915-5G
BT219915-25GEA 405 USD
BT219915-25G BT219915-5G
Tin(II) iodide, anhydrous ≥99.999% (trace metals basis)
Tin(II) iodide

CAS: 10294-70-9; EC No: 233-667-3; MDL No: MFCD00049545 UN No: UN3260; Haz Class: 8; Packing Group: III -10 Mesh Beads; Linear Formula: SnI2; MW: 372.52 Melting Point: 320°; Boiling Point: 714° (decomposes) Density (g/mL): 5.28


  • -10 Mesh Beads

Formula: I₂Sn
MW: 372.52 g/mol
Boiling Pt: 714 °C (dec.)
Melting Pt: 320 °C
Density: 5.28 g/mL
MDL Number: MFCD00049545
CAS Number: 10294-70-9
EINECS: 233-667-3
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR