N,N-Dimethyl-1-naphthylamine ≥99%
Supplier: BeanTown Chemical
Synonyms:
Dimethyl(1-naphthyl)amine, N,N-Dimethylnaphthalen-1-amine
219895-25ML
219895-5ML
BT219895-25MLEA
77.57
USD
BT219895-25ML
BT219895-5ML
N,N-Dimethyl-1-naphthylamine ≥99%
N,N-Dimethyl-1-naphthylamine
CAS: 86-56-6; EC No: 201-682-4; MDL No: MFCD00003919
UN No: UN3082; Haz Class: 9; Packing Group: III
Liquid; Linear Formula: C10H7N(CH3)2; Molecular Formula: C12H13N; MW: 171.24
Boiling Point: 139-140°/13 mmHg; Flash point:
- Refractive index: 1.622
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₁₂H₁₃N MW: 171.24 g/mol Boiling Pt: 139…140 °C/13 mmHg Density: 1.042 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00003919 CAS Number: 86-56-6 EINECS: 201-682-4 UN: 3082 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...