N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-(Chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane Bis(tetrafluoroborate), F-TEDA-BF4, 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

219865-25G 219865-5G
BT219865-25GEA 163 USD
BT219865-25G BT219865-5G
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥98%
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)

CAS: 140681-55-6; EC No: 414-380-4; MDL No: MFCD00142607 UN No: UN3088; Haz Class: 4.2; Packing Group: II Powder; Molecular Formula: C7H14B2ClF9N2; MW: 354.26 Melting Point: 260°


  • Powder

Formula: C₇H₁₄B₂ClF₉N₂
Melting Pt: 260 °C
MDL Number: MFCD00142607
CAS Number: 140681-55-6
EINECS: 414-380-4
ADR: 4.2,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR