1,1'-Carbonyldiimidazole ≥97%

Supplier: BeanTown Chemical

Synonyms: Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI

219820-10G 219820-250G 219820-50G
BT219820-10GEA 54.83 USD
BT219820-10G BT219820-250G BT219820-50G
1,1'-Carbonyldiimidazole ≥97%
1,1'-Carbonyldiimidazole

CAS: 530-62-1; EC No: 208-488-9; MDL No: MFCD00005286 UN No: UN3263; Haz Class: 8; Packing Group: II Solid; Molecular Formula: C7H6N4O; MW: 162.15 Melting Point: 117-122° Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: C₇H₆N₄O
MW: 162.15 g/mol
Melting Pt: 117…122 °C
MDL Number: MFCD00005286
CAS Number: 530-62-1
EINECS: 208-488-9
UN: 3263
ADR: 8 II
Merck Index: 14,01819

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR