Diethyl cyanophosphonate ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Diethyl phosphorocyanidate
219815-25G
219815-5G
BT219815-25GEA
131
USD
BT219815-25G
BT219815-5G
Diethyl cyanophosphonate ≥90%, Technical Grade
Diethyl cyanophosphonate
CAS: 2942-58-7; EC No: 220-936-5; MDL No: MFCD00010256; RTECS: TD2500000
UN No: UN2922; Haz Class: 8 (6.1); Packing Group: II
Liquid; Molecular Formula: C5H10NO3P; MW: 163.11
Boiling Point: 104-105°/19 mmHg; Flash point: 80°C (176°F)
Density (g/mL): 1.075; Refractive Index: 1.403
Moisture Sensitive
- RTECS: TD2500000
- Refractive index: 1.403
- Moisture Sensitive
- Liquid
Formula:
C₅H₁₀NO₃P Boiling Pt: 104…105 °C/19 mmHg Density: 1.075 g/mL Flash Pt: 80 °C (176 °F) |
MDL Number:
MFCD00010256 CAS Number: 2942-58-7 EINECS: 220-936-5 ADR: 8 (6.1),II |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...