Thiophene ≥99%
Supplier: BeanTown Chemical
219750-250G
219750-50G
BT219750-250GEA
32
USD
BT219750-250G
BT219750-50G
Thiophene ≥99%
Thiophene
CAS: 110-02-1; EC No: 203-729-4; MDL No: MFCD00005413
UN No: UN2414; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C4H4S; MW: 84.14
Melting Point: -38°; Boiling Point: 83-85°; Flash point: -9°C (16°F)
Density (g/mL): 1.051; Refractive Index: 1.529
- Refractive index: 1.529
- Liquid
Formula:
C₄H₄S MW: 84.14 g/mol Boiling Pt: 83…85 °C Melting Pt: –38 °C Density: 1.051 g/mL Flash Pt: –9 °C (16 °F) |
MDL Number:
MFCD00005413 CAS Number: 110-02-1 EINECS: 203-729-4 UN: 2414 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...