Silver(I) oxide
Supplier: BeanTown Chemical
Synonyms:
di-Silver oxide, Disilver oxide, Silver oxide
219735-10G
219735-250G
219735-50G
BT219735-10GEA
67.52
USD
BT219735-10G
BT219735-250G
BT219735-50G
Silver(I) oxide
Silver(I) oxide
CAS: 20667-12-3; EC No: 243-957-1; MDL No: MFCD00003404; RTECS: VW4900000
UN No: UN1479; Haz Class: 5.1; Packing Group: II
-325 Mesh Powder; Molecular Formula: Ag2O; MW: 231.74
Melting Point: 230° (decomposes)
Density (g/mL): 7.143
Light Sensitive
- RTECS: VW4900000
- -325 Mesh Powder
- Light Sensitive
Formula:
Ag₂O MW: 231.74 g/mol Melting Pt: 230 °C (dec.) Density: 7.143 g/mL |
MDL Number:
MFCD00003404 CAS Number: 20667-12-3 UN: 1479 ADR: 5.1 II Merck Index: 12,08665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...