3-Cyclopentylpropionic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-Cyclopentylpropanoic acid, 3-Cylcopentylpropanoic acid
219725-100G
219725-25G
BT219725-100GEA
193
USD
BT219725-100G
BT219725-25G
3-Cyclopentylpropionic acid ≥98%
3-Cyclopentylpropionic acid
CAS: 140-77-2; EC No: 205-433-0; MDL No: MFCD00001392
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C8H14O2; MW: 142.20
Boiling Point: 130-132°/12 mmHg; Flash point: 47°C (117°F)
Density (g/mL): 0.996; Refractive Index: 1.457
- Refractive index: 1.457
- Liquid
Formula:
C₈H₁₄O₂ MW: 142.2 g/mol Boiling Pt: 130…132 °C/12 mmHg Density: 0.996 g/mL Flash Pt: 47 °C (117 °F) |
MDL Number:
MFCD00001392 CAS Number: 140-77-2 EINECS: 205-433-0 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...