Trisodium phosphate dodecahydrate 98.0-102.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate
219695-2KG
219695-500G
BT219695-2KGEA
176
USD
BT219695-2KG
BT219695-500G
Trisodium phosphate dodecahydrate 98.0-102.0% ACS
Trisodium phosphate dodecahydrate
CAS: 10101-89-0; EC No: 231-509-8; MDL No: MFCD00149198; RTECS: TC9575000
UN No: UN3262; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: Na3PO4·12H2O; MW: 380.12
Density (g/mL): 1.62
- RTECS: TC9575000
- Crystalline
Formula:
Na₃PO₄·12H₂O MW: 380.12 g/mol Melting Pt: 75 Density: 1.62 g/mL |
MDL Number:
MFCD00149198 CAS Number: 10101-89-0 EINECS: 231-509-8 ADR: 8 III Merck Index: 13,08736 |
Specification Test Results
Assay | 98.0-102.0% |
Excess alkali | ≤2.5% |
Insoluble matter | ≤0.01% |
Chloride (Cl) | ≤0.001% |
Sulfate (SO4) | ≤0.01% |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤3ppm |
Nitrogen compounds | ≤0.001% |
Clarity of solution | Clear and colorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...