Tetrabutylammonium fluoride trihydrate ≥98%
Supplier: BeanTown Chemical
219690-10G
219690-50G
BT219690-10GEA
58
USD
BT219690-10G
BT219690-50G
Tetrabutylammonium fluoride trihydrate ≥98%
Tetrabutylammonium fluoride trihydrate
CAS: 87749-50-6; EC No: 207-057-2; MDL No: MFCD00149981
UN No: UN1759; Haz Class: 8; Packing Group: III
Crystalline/Powder; Linear Formula: [CH3(CH2)3]4N(F)·3H2O; Molecular Formula: C16H36FN·3H2O; MW: 315.52
Melting Point: 62-63°
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
C₁₆H₃₆FN·3H₂O MW: 315.51 g/mol Melting Pt: 62…63 °C |
MDL Number:
MFCD00149981 CAS Number: 87749-50-6 EINECS: 207-057-2 UN: 1759 ADR: 6.1 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...