Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
Supplier: BeanTown Chemical
Synonyms:
Bis(norbornadiene)rhodium(I) tetrafluoroborate, Bis(norbornadiene)rhodium (I) tetrafluoroborate
219640-1G
219640-5G
BT219640-1GEA
334
USD
BT219640-1G
BT219640-5G
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate
CAS: 36620-11-8; MDL No: MFCD00671775
UN No: UN1759; Haz Class: 8; Packing Group: II
Powder; Linear Formula: [Rh(C7H8)2]BF4; Molecular Formula: C14H16BF4Rh; MW: 373.99
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
C₁₄H₈BF₄Rh MW: 365.92 g/mol |
MDL Number:
MFCD00671775 CAS Number: 36620-11-8 ADR: 8,II |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...