Calcium, granules -9 mesh
Supplier: BeanTown Chemical
219630-1KG
219630-250G
219630-50G
BT219630-1KGEA
240.08
USD
BT219630-1KG
BT219630-250G
BT219630-50G
Calcium, granules -9 mesh
Calcium
CAS: 7440-70-2; EC No: 213-179-5; MDL No: MFCD00085314; RTECS: EV8040000
UN No: UN1401; Haz Class: 4.3; Packing Group: II
Solid; Molecular Formula: Ca; MW: 40.08
Melting Point: 850°; Boiling Point: 1484°
Density (g/mL): 1.54
- RTECS: EV8040000
- Solid
Formula:
Ca MW: 40.08 g/mol Boiling Pt: 1484 °C Melting Pt: 850 °C Density: 1.54 g/mL |
MDL Number:
MFCD00085314 CAS Number: 7440-70-2 EINECS: 213-179-5 UN: 1401 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...