Tetrabutylammonium fluoride 1 M in THF
Supplier: BeanTown Chemical
219625-100ML
219625-25ML
219625-500ML
BT219625-100MLEA
66.82
USD
BT219625-100ML
BT219625-25ML
BT219625-500ML
Tetrabutylammonium fluoride 1 M in THF
Tetrabutylammonium fluoride
CAS: 429-41-4; EC No: 207-057-2; MDL No: MFCD00011747
UN No: UN2924; Haz Class: 3(8); Packing Group: II
Liquid; Linear Formula: [CH3(CH2)3]4N(F); Molecular Formula: C16H36FN; MW: 261.47
Flash point: -17°C (1°F)
Density (g/mL): 0.903
Hygroscopic
- Hygroscopic
- Liquid
Formula:
C₁₆H₃₆FN MW: 261.47 g/mol Density: 0.903 g/mL Flash Pt: –17 °C (1 °F) |
MDL Number:
MFCD00011747 CAS Number: 429-41-4 EINECS: 207-057-2 UN: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...