Titanium(IV) oxide sulfate sulfuric acid hydrate
Supplier: BeanTown Chemical
Synonyms:
Titanyl sulphate, Titanium(IV) oxysulphate-sulphuric acid
219615-2KG
219615-500G
BT219615-2KGEA
265
USD
BT219615-2KG
BT219615-500G
Titanium(IV) oxide sulfate sulfuric acid hydrate
Titanium(IV) oxide sulfate sulfuric acid hydrate
CAS: 123334-00-9; EC No: 237-523-0; MDL No: MFCD00011270
UN No: UN3260; Haz Class: 8; Packing Group: II
-10 Mesh Powder; Linear Formula: TiOSO4·xH2O + H2SO4
- -10 Mesh Powder
Formula:
TiOSO₄·xH₂SO₄·yH₂O |
MDL Number:
MFCD00150565 CAS Number: 123334-00-9 EINECS: 237-523-0 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...