(Chloromethyl)trimethylsilane ≥98%

Supplier: BeanTown Chemical

219595-10G 219595-250G 219595-50G
BT219595-10GEA 49 USD
BT219595-10G BT219595-250G BT219595-50G
(Chloromethyl)trimethylsilane ≥98%
(Chloromethyl)trimethylsilane

CAS: 2344-80-1; EC No: 219-058-5; MDL No: MFCD00000878 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3SiCH2Cl; Molecular Formula: C4H11ClSi; MW: 122.67 Boiling Point: 98-99°; Flash point: -4°C (25°F) Density (g/mL): 0.879; Refractive Index: 1.418 Moisture Sensitive


  • Refractive index: 1.418
  • Moisture Sensitive
  • Liquid

Formula: ClH₂CSi(CH₃)₃
Boiling Pt: 98…99 °C
Density: 0.879 g/mL
Flash Pt: –4 °C (25 °F)
MDL Number: MFCD00000878
CAS Number: 2344-80-1
EINECS: 219-058-5
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR