Tetrabutylammonium perchlorate
Supplier: BeanTown Chemical
Synonyms:
Tetra-n-butylammonium perchlorate
219580-10G
219580-50G
BT219580-10GEA
47.07
USD
BT219580-10G
BT219580-50G
Tetrabutylammonium perchlorate
Tetrabutylammonium perchlorate
CAS: 1923-70-2; EC No: 217-655-5; MDL No: MFCD00038722
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline/Powder; Linear Formula: [CH3(CH2)3]4N(ClO4); Molecular Formula: C16H36ClNO4 ; MW: 341.92
Melting Point: 210-215°
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
C₁₆H₃₆ClNO₄ MW: 341.92 g/mol Melting Pt: 210…215 °C |
MDL Number:
MFCD00038722 CAS Number: 1923-70-2 EINECS: 217-655-5 UN: 1481 ADR: 5.1,II |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...