4-Aminopyridine ≥98%

Supplier: BeanTown Chemical

219575-1KG 219575-250G 219575-50G
BT219575-1KGEA 790 USD
BT219575-1KG BT219575-250G BT219575-50G
4-Aminopyridine ≥98%
4-Aminopyridine

CAS: 504-24-5; EC No: 207-987-9; MDL No: MFCD00006439; RTECS: US1750000 UN No: UN2671; Haz Class: 6.1; Packing Group: II Crystalline; Molecular Formula: C5H6N2; MW: 94.11 Melting Point: 155-158°; Boiling Point: 273°


  • RTECS: US1750000
  • Crystalline

Formula: C₅H₆N₂
Boiling Pt: 273 °C
Melting Pt: 155…158 °C
MDL Number: MFCD00006439
CAS Number: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR