1,3-Divinyltetramethyldisiloxane ≥96% (max. 4% 1-vinyl-3-ethyltetramethyldisiloxane)
Supplier: BeanTown Chemical
CAS: 2627-95-4; EC No: 220-099-6; MDL No: MFCD00014933; RTECS: JM9235250
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C8H18OSi2; MW: 186.40
Melting Point: -99°; Boiling Point: 139°; Flash point: 19°C (66°F)
Density (g/mL): 0.809; Refractive Index: 1.411
Moisture Sensitive
- RTECS: JM9235250
- Refractive index: 1.411
- Moisture Sensitive
- Liquid
Formula:
C₈H₁₈OSi₂ Boiling Pt: 139 °C Melting Pt: –99 °C Density: 0.809 g/mL Flash Pt: 19 °C (66 °F) |
MDL Number:
MFCD00014933 CAS Number: 2627-95-4 EINECS: 220-099-6 ADR: 3,III |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...