Silver perchlorate, anhydrous
Supplier: BeanTown Chemical
219490-100G
219490-25G
219490-5G
BT219490-100GEA
675
USD
BT219490-100G
BT219490-25G
BT219490-5G
Silver perchlorate, anhydrous
Silver perchlorate
CAS: 7783-93-9; EC No: 232-035-4; MDL No: MFCD00003400
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline; Molecular Formula: AgClO4; MW: 207.32
Melting Point: 485° (decomposes)
Density (g/mL): 2.806
Hygroscopic , Light Sensitive
- Hygroscopic, Light Sensitive
- Crystalline
Formula:
AgClO₄ Melting Pt: 485 °C (dec.) Density: 2.806 g/mL |
MDL Number:
MFCD00003400 CAS Number: 7783-93-9 EINECS: 232-035-4 UN: 1481 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...