Dichlorophenylborane ≥96%

Supplier: BeanTown Chemical

Synonyms: Phenylboron dichloride

219450-1G 219450-25G 219450-5G
BT219450-1GEA 27.27 USD
BT219450-1G BT219450-25G BT219450-5G
Dichlorophenylborane ≥96%
Dichlorophenylborane

CAS: 873-51-8; MDL No: MFCD00013612 UN No: UN2924; Haz Class: 3(8); Packing Group: II Liquid; Molecular Formula: C6H5BCl2; MW: 158.83 Boiling Point: 66°/11 mmHg; Flash point: 14°C (57°F) Density (g/mL): 1.224; Refractive Index: 1.545 Moisture Sensitive


  • Refractive index: 1.545
  • Moisture Sensitive
  • Liquid

Formula: C₆H₅BCl₂
MW: 158.82 g/mol
Boiling Pt: 66 °C/11 mmHg
Density: 1.224 g/mL
Flash Pt: 14 °C (57 °F)
MDL Number: MFCD00013612
CAS Number: 873-51-8
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR