(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) ≥98%
Supplier: BeanTown Chemical
Synonyms:
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
219445-1G
219445-25G
219445-5G
BT219445-1GEA
21
USD
BT219445-1G
BT219445-25G
BT219445-5G
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) ≥98%
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
CAS: 56602-33-6; EC No: 260-279-1; MDL No: MFCD00011948
UN No: UN1325; Haz Class: 4.1; Packing Group: II
Powder; Molecular Formula: C12H22F6N6OP2; MW: 442.28
Melting Point: 130° (decomposes)
Light Sensitive, Moisture Sensitive
- Light Sensitive, Moisture Sensitive
- Powder
Formula:
C₁₂H₂₂F₆N₆OP₂ MW: 442.28 g/mol Melting Pt: 130 °C (dec.) |
MDL Number:
MFCD00011948 CAS Number: 56602-33-6 EINECS: 260-279-1 ADR: 4.1 II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...