Zinc oxide 99,9995

Supplier: BeanTown Chemical

Synonyms: Zinci oxidum, Zinc (II) oxide

219425-100G 219425-25G
BT219425-100GEA 397.56 USD
BT219425-100G BT219425-25G
Zinc oxide 99,9995
Zinc oxide

CAS: 1314-13-2; EC No: 215-222-5; MDL No: MFCD00011300; RTECS: ZH4810000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: ZnO; MW: 81.39 Melting Point: 1975° Density (g/mL): 5.61


  • RTECS: ZH4810000
  • Powder

Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.61 g/mL
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Assay 99.9995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR