Cadmium(II) chloride, anhydrous ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Cadmium chloride
219320-100G
219320-500G
BT219320-100GEA
97.09
USD
BT219320-100G
BT219320-500G
Cadmium(II) chloride, anhydrous ≥99.0% ACS
Cadmium(II) chloride
CAS: 10108-64-2; EC No: 233-296-7; MDL No: MFCD00010916; RTECS: EV0175000
UN No: UN2570; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: CdCl2; MW: 183.32
Melting Point: 568°
Hygroscopic
- RTECS: EV0175000
- Hygroscopic
- Powder
Formula:
CdCl₂ MW: 183.32 g/mol Melting Pt: 568 °C |
MDL Number:
MFCD00010916 CAS Number: 10108-64-2 EINECS: 233-296-7 UN: 2570 ADR: 6.1,III |
Specification Test Results
Assay | 99.0% min. |
Insoluble matter | ≤0.01% |
Ammonium (NH4) | ≤0.01% |
Nitrate & nitrite | ≤0.003% |
Sulfate (SO4) | ≤0.01% |
Iron (Fe) | ≤10ppm |
Copper (Cu) | ≤10ppm |
Lead (Pb) | ≤50ppm |
Zinc (Zn) | ≤500ppm |
Sodium (Na) | ≤500ppm |
Potassium (K) | ≤200ppm |
Calcium (Ca) | ≤100ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...