4-Chloro-3-(trifluoromethoxy)aniline ≥97%

Supplier: BeanTown Chemical

219305-1G 219305-250MG
BT219305-1GEA 288 USD
BT219305-1G BT219305-250MG
4-Chloro-3-(trifluoromethoxy)aniline ≥97%
4-Chloro-3-(trifluoromethoxy)aniline

CAS: 97608-50-9; MDL No: MFCD09832255 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C7H5ClF3NO; MW: 211.57 Refractive Index: 1.495


  • Refractive index: 1.495
  • Liquid

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD09832255
CAS Number: 97608-50-9
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR