Pentafluorophenol ≥99%
Supplier: BeanTown Chemical
Synonyms:
2,3,4,5,6-Pentafluorophenol
219285-100G
219285-25G
219285-5G
BT219285-100GEA
388
USD
BT219285-100G
BT219285-25G
BT219285-5G
Pentafluorophenol ≥99%
Pentafluorophenol
CAS: 771-61-9; EC No: 212-235-8; MDL No: MFCD00002156; RTECS: SM6680000
UN No: UN3261; Haz Class: 8; Packing Group: II
Solid; Linear Formula: C6F5OH; MW: 184.06
Melting Point: 34-36°; Boiling Point: 143°; Flash point: 72°C (161°F)
Density (g/mL): 1.757; Refractive Index: 1.427
Moisture Sensitive
- RTECS: SM6680000
- Refractive index: 1.427
- Moisture Sensitive
- Solid
Formula:
C₆HF₅O MW: 184.07 g/mol Boiling Pt: 143 °C Melting Pt: 34…36 °C Density: 1.757 g/mL Flash Pt: 72 °C (161 °F) |
MDL Number:
MFCD00002156 CAS Number: 771-61-9 EINECS: 212-235-8 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...