Piperidine ≥99%
Supplier: BeanTown Chemical
Synonyms:
Hexahydropyridine, Hexazane, Pentamethyleneimine
219260-100ML
219260-2.5L
219260-500ML
BT219260-100MLEA
33.4
USD
BT219260-100ML
BT219260-2.5L
BT219260-500ML
Piperidine ≥99%
Piperidine
CAS: 110-89-4; EC No: 203-813-0; MDL No: MFCD00005979; RTECS: TM3500000
UN No: UN2401; Haz Class: 8 (3); Packing Group: I
Liquid; Molecular Formula: C5H11N; MW: 85.15
Melting Point: -13°; Boiling Point: 105-106°; Flash point: 3°C (37°F)
Density (g/mL): 0.862; Refractive Index: 1.452
Air Sensitive
- RTECS: TM3500000
- Refractive index: 1.452
- Air Sensitive
- Liquid
Formula:
C₅H₁₁N MW: 85.15 g/mol Boiling Pt: 105…106 °C Melting Pt: –13 °C Density: 0.862 g/mL Flash Pt: 3 °C (37 °F) |
MDL Number:
MFCD00005979 CAS Number: 110-89-4 EINECS: 203-813-0 UN: 2401 ADR: 8,I |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...