(±)-2-Bromoisovaleric acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Bromo-3-methylbutyricacid, 2-Bromo-3-methylbutyric acid
219210-10G
219210-50G
BT219210-10GEA
33
USD
BT219210-10G
BT219210-50G
(±)-2-Bromoisovaleric acid ≥97%
(±)-2-Bromoisovaleric acid
CAS: 565-74-2; EC No: 209-291-0; MDL No: MFCD00004210; RTECS: ES8055000
UN No: UN3261; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: C5H9BrO2 ; MW: 181.03
Melting Point: 39-42°; Boiling Point: 124-126°/20 mmHg; Flash point: 107°C (225°F)
- RTECS: ES8055000
- Crystalline
Formula:
C₅H₉BrO₂ MW: 181.03 g/mol Boiling Pt: 124…126 °C/20 mmHg Melting Pt: 39…42 °C Flash Pt: 107 °C (225 °F) |
MDL Number:
MFCD00004210 CAS Number: 565-74-2 EINECS: 209-291-0 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...