1-Methylpiperazine ≥98%
Supplier: BeanTown Chemical
219115-100ML
219115-2.5L
219115-500ML
BT219115-100MLEA
26
USD
BT219115-100ML
BT219115-2.5L
BT219115-500ML
1-Methylpiperazine ≥98%
1-Methylpiperazine
CAS: 109-01-3; EC No: 203-639-5; MDL No: MFCD00005966; RTECS: TM1225000
UN No: UN2734; Haz Class: 8 (3); Packing Group: II
Liquid; Molecular Formula: C5H12N2 ; MW: 100.16
Boiling Point: 138°; Flash point: 39°C (102°F)
Density (g/mL): 0.903; Refractive Index: 1.466
Air Sensitive
- RTECS: TM1225000
- Refractive index: 1.466
- Air Sensitive
- Liquid
Formula:
C₅H₁₂N₂ MW: 100.16 g/mol Boiling Pt: 138 °C Density: 0.903 g/mL Flash Pt: 39 °C (102 °F) |
MDL Number:
MFCD00005966 CAS Number: 109-01-3 EINECS: 203-639-5 UN: 2734 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...