Isobutyl isobutyrate ≥98%
Supplier: BeanTown Chemical
219105-1L
219105-250ML
BT219105-1LEA
100
USD
BT219105-1L
BT219105-250ML
Isobutyl isobutyrate ≥98%
Isobutyl isobutyrate
CAS: 97-85-8; EC No: 202-612-5; MDL No: MFCD00008916; RTECS: NQ5250000
UN No: UN2528; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: (CH3)2CHCOOCH2CH(CH3)2 ; Molecular Formula: C8H16O2; MW: 144.22
Melting Point: -81°; Boiling Point: 145-152°; Flash point: 37°C (98°F)
Density (g/mL): 0.855; Refractive Index: 1.398
- RTECS: NQ5250000
- Refractive index: 1.398
- Liquid
Formula:
C₈H₁₆O₂ MW: 144.21 g/mol Boiling Pt: 145…152 °C Melting Pt: –81 °C Density: 0.855 g/mL Flash Pt: 37 °C (98 °F) |
MDL Number:
MFCD00008916 CAS Number: 97-85-8 EINECS: 202-612-5 UN: 2528 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...