2,3,5,6-Tetrafluorophenol ≥97%

Supplier: BeanTown Chemical

219025-25G 219025-5G
BT219025-25GEA 178.47 USD
BT219025-25G BT219025-5G
2,3,5,6-Tetrafluorophenol ≥97%
2,3,5,6-Tetrafluorophenol

CAS: 769-39-1; EC No: 212-209-6; MDL No: MFCD00002157 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Molecular Formula: C6H2F4O; MW: 166.07 Melting Point: 37-39°; Boiling Point: 140°; Flash point: 79°C (174°F)


  • Solid

Formula: C₆H₂F₄O
MW: 166.07 g/mol
Boiling Pt: 140 °C
Melting Pt: 37…39 °C
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00002157
CAS Number: 769-39-1
EINECS: 212-209-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR