Potassium fluoride, anhydrous ≥99%

Supplier: BeanTown Chemical

219020-1KG 219020-250G 219020-50G
BT219020-1KGEA 274.12 USD
BT219020-1KG BT219020-250G BT219020-50G
Potassium fluoride, anhydrous ≥99%
Potassium fluoride

CAS: 7789-23-3; EC No: 232-151-5; MDL No: MFCD00011398; RTECS: TT0700000 UN No: UN1812; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: KF; MW: 58.1 Melting Point: 858°; Boiling Point: 1505° Density (g/mL): 2.48; Refractive Index: 1.363 Hygroscopic


  • RTECS: TT0700000
  • Refractive index: 1.363
  • Hygroscopic
  • Powder

Formula: KF
MW: 58.1 g/mol
Boiling Pt: 1505 °C
Melting Pt: 858 °C
Density: 2.48 g/mL
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR