Cobalt(III) fluoride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Cobaltic fluoride, Cobalt trifluoride
219015-100G
219015-25G
BT219015-100GEA
227
USD
BT219015-100G
BT219015-25G
Cobalt(III) fluoride ≥99%
Cobalt(III) fluoride
CAS: 10026-18-3; EC No: 233-062-4; MDL No: MFCD00010942
UN No: UN3085; Haz Class: 5.1 (8); Packing Group: III
Powder; Molecular Formula: CoF3; MW: 115.93
Density (g/mL): 3.88
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
CoF₃ MW: 115.93 g/mol Density: 3.88 g/mL |
MDL Number:
MFCD00010942 CAS Number: 10026-18-3 EINECS: 233-062-4 UN: 3085 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...