2-Butyne-1-ol ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Butyn-1-ol, But-2-yn-1-ol, 1-Hydroxy-2-butyne
218955-100G
218955-25G
218955-5G
BT218955-100GEA
475
USD
BT218955-100G
BT218955-25G
BT218955-5G
2-Butyne-1-ol ≥98%
2-Butyne-1-ol
CAS: 764-01-2; EC No: 212-113-4; MDL No: MFCD00002914
UN No: UN1987; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3C≡CCH2OH; Molecular Formula: C4H6O; MW: 70.09
Melting Point: -2°; Boiling Point: 142-143°; Flash point: 51°C (124°F)
Density (g/mL): 0.937; Refractive Index: 1.453
- Refractive index: 1.453
- Liquid
Formula:
C₄H₆O MW: 70.09 g/mol Boiling Pt: 142…143 °C Melting Pt: –2 °C Density: 0.937 g/mL Flash Pt: 51 °C (124 °F) |
MDL Number:
MFCD00002914 CAS Number: 764-01-2 EINECS: 212-113-4 UN: 1987 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...