Cerium ≥99.9% (trace metals basis excluding Ta), rods Ø 6.35 mm
Supplier: BeanTown Chemical
218945-100MM
218945-10MM
218945-50MM
BT218945-100MMEA
626
USD
BT218945-100MM
BT218945-10MM
BT218945-50MM
Cerium ≥99.9% (trace metals basis excluding Ta), rods Ø 6.35 mm
Cerium
CAS: 7440-45-1; EC No: 231-154-9; MDL No: MFCD00010924
UN No: UN1333; Haz Class: 4.1; Packing Group: II
Solid, cast rod, diameter is approximate; Molecular Formula: Ce; MW: 140.12
Melting Point: 795°; Boiling Point: 3443°
Density (g/mL): 6.67
- Solid, cast rod, diameter is approximate
Formula:
Ce MW: 140.12 g/mol Boiling Pt: 3468 Melting Pt: 795 Density: 6.67 g/mL |
MDL Number:
MFCD00010924 CAS Number: 7440-45-1 EINECS: 231-154-9 UN: 1333 ADR: 4.1 II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...