Molybdenum(VI) oxide ≥99.95% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Molybdenum trioxide

218915-100G 218915-2KG 218915-500G
BT218915-100GEA 58 USD
BT218915-100G BT218915-2KG BT218915-500G
Molybdenum(VI) oxide ≥99.95% (trace metals basis)
Molybdenum(VI) oxide

CAS: 1313-27-5; EC No: 215-204-7; MDL No: MFCD00003469; RTECS: QA4725000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Sublimed Powder; Molecular Formula: MoO3; MW: 143.94 Melting Point: 795°; Boiling Point: 1155° (sublimes) Density (g/mL): 4.692


  • RTECS: QA4725000
  • Sublimed Powder

Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1155 °C (subl.)
Melting Pt: 795 °C
Density: 4.692 g/mL
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR