Methanol ≥99.8% for HPLC

Supplier: BeanTown Chemical

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

218905-1L 218905-4L 218905-4X4L
BT218905-1LEA 45.43 USD
BT218905-1L BT218905-4L BT218905-4X4L
Methanol ≥99.8% for HPLC
Methanol

CAS: 67-56-1; EC No: 200-659-6; MDL No: MFCD00004595; RTECS: PC1400000 UN No: UN1230; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3OH; MW: 32.04 Melting Point: -98°; Boiling Point: 64.7°; Flash point: 9.7°C (49.5°F) Density (g/mL): 0.791; Refractive Index: 1.329


  • RTECS: PC1400000
  • Refractive index: 1.329
  • Liquid

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.791 g/mL
Flash Pt: 9.7 °C (49.5 °F)
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.8% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR