4-Bromo-3-methylbenzenesulfonyl chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Bromo-3-methylbenzene-1-sulphonyl chloride
218900-1G
218900-5G
BT218900-1GEA
57
USD
BT218900-1G
BT218900-5G
4-Bromo-3-methylbenzenesulfonyl chloride ≥97%
4-Bromo-3-methylbenzenesulphonyl chloride
CAS: 72256-93-0; MDL No: MFCD03094696
UN No: UN3261; Haz Class: 8; Packing Group: II
Solid; Molecular Formula: C7H6BrClO2S ; MW: 269.54
Melting Point: 58-61°
Moisture Sensitive
- Moisture Sensitive
- Solid
Formula:
C₇H₆BrClO₂S MW: 269.55 g/mol Melting Pt: 58…61 °C |
MDL Number:
MFCD03094696 CAS Number: 72256-93-0 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...