Phosphonoacetic acid ≥98%

Supplier: BeanTown Chemical

218880-100G 218880-10G 218880-25G
BT218880-100GEA 417 USD
BT218880-100G BT218880-10G BT218880-25G
Phosphonoacetic acid ≥98%
Phosphonoacetic acid

CAS: 4408-78-0; EC No: 224-558-1; MDL No: MFCD00004311; RTECS: AJ3278000 UN No: UN3261; Haz Class: 8; Packing Group: III Solid; Linear Formula: (HO)2P(O)CH2COOH; Molecular Formula: C2H5O5P; MW: 140.04 Melting Point: 143-146° Hygroscopic


  • RTECS: AJ3278000
  • Hygroscopic
  • Solid

Formula: C₂H₅O₅P
MW: 140.04 g/mol
Melting Pt: 143…146 °C
MDL Number: MFCD00004311
CAS Number: 4408-78-0
EINECS: 224-558-1
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR