Barium titanate ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
218840-100G
218840-500G
BT218840-100GEA
79.68
USD
BT218840-100G
BT218840-500G
Barium titanate ≥99.5% (trace metals basis)
Barium titanate
CAS: 12047-27-7; EC No: 234-975-0; MDL No: MFCD00003447
UN No: UN1564; Haz Class: 6.1; Packing Group: III
- <2 Micron Powder
Formula:
BaO₃Ti MW: 233.21 g/mol Density: 6.08 g/mL |
MDL Number:
MFCD00003447 CAS Number: 12047-27-7 EINECS: 234-975-0 UN: 1564 ADR: 6.1,III |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...