Cobalt(II) nitrate hexahydrate ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate

218765-10G 218765-50G
BT218765-10GEA 113.45 USD
BT218765-10G BT218765-50G
Cobalt(II) nitrate hexahydrate ≥99.999%
Cobalt(II) nitrate hexahydrate

CAS: 10026-22-9; EC No: 233-402-1; MDL No: MFCD00149647; RTECS: QU7355500 UN No: UN1477; Haz Class: 5.1; Packing Group: III Wet Crystalline Aggregates; Linear Formula: Co(NO3)2·6H2O; MW: 291.04 Melting Point: 55°


  • RTECS: QU7355500
  • Wet Crystalline Aggregates

Formula: Co(NO₃)₂·6H₂O
Melting Pt: 55 °C
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR