3,4-Diaminopyridine ≥98%
Supplier: BeanTown Chemical
Synonyms:
3,4-Pyridinediamine, Amifampridine, 3,4-DAP, Pyridine-3,4-diyldiamine, Pyridine-3,4-diamine
218670-1G
218670-25G
218670-5G
BT218670-1GEA
38
USD
BT218670-1G
BT218670-25G
BT218670-5G
3,4-Diaminopyridine ≥98%
3,4-Diaminopyridine
CAS: 54-96-6; EC No: 200-220-9; MDL No: MFCD00006401; RTECS: US7600000
UN No: UN2811; Haz Class: 6.1; Packing Group: II
Crystalline; Molecular Formula: C5H7N3 ; MW: 109.13
Melting Point: 216-218°
- RTECS: US7600000
- Crystalline
Formula:
C₅H₇N₃ MW: 109.13 g/mol Melting Pt: 216…218 °C |
MDL Number:
MFCD00006401 CAS Number: 54-96-6 EINECS: 200-220-9 UN: 2811 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...